Product Details

SNP ID
rs10270436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149884243 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAATCAGCTGGCAGGCACCACAG[G/T]TTTGCTTGACTTGTAGCACTGGAAG
Phenotype
MIM: 611019
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACTR3C PubMed Links

Gene Details

Gene
ACTR3C
Gene Name
ARP3 actin-related protein 3 homolog C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164458.1 4567 Intron NP_001157930.1
NM_001164459.1 4567 Intron NP_001157931.1
XM_005250043.4 4567 Intron XP_005250100.3
XM_011516502.2 4567 Intron XP_011514804.1
XM_011516506.2 4567 Intron XP_011514808.1
XM_011516507.2 4567 Intron XP_011514809.1
XM_011516508.2 4567 Intron XP_011514810.1
XM_011516511.2 4567 Intron XP_011514813.1
XM_017012548.1 4567 UTR 3 XP_016868037.1
XM_017012549.1 4567 Intron XP_016868038.1
XM_017012550.1 4567 UTR 3 XP_016868039.1
XM_017012551.1 4567 UTR 3 XP_016868040.1
XM_017012552.1 4567 Intron XP_016868041.1
XM_017012553.1 4567 Intron XP_016868042.1
Gene
ATP6V0E2
Gene Name
ATPase H+ transporting V0 subunit e2
There are no transcripts associated with this gene.

View Full Product Details