Product Details

SNP ID
rs9511434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:24749510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAAACAATTGCTTAAGATTTTA[A/G]AATCAATTTCCCTTATTCTTTTTCT
Phenotype
MIM: 605793
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF17 PubMed Links

Gene Details

Gene
RNF17
Gene Name
ring finger protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184993.1 Intron NP_001171922.1
NM_031277.2 Intron NP_112567.2
XM_006719846.3 Intron XP_006719909.1
XM_006719849.2 Intron XP_006719912.1
XM_006719852.2 Intron XP_006719915.1
XM_011535152.2 Intron XP_011533454.1
XM_011535155.2 Intron XP_011533457.1
XM_011535156.2 Intron XP_011533458.1
XM_011535157.2 Intron XP_011533459.1
XM_011535158.2 Intron XP_011533460.1
XM_011535159.2 Intron XP_011533461.1
XM_011535160.2 Intron XP_011533462.1
XM_011535162.1 Intron XP_011533464.1
XM_011535163.1 Intron XP_011533465.1
XM_011535164.2 Intron XP_011533466.1
XM_017020676.1 Intron XP_016876165.1

View Full Product Details