Product Details

SNP ID
rs6009672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:49415737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTTTTACAGCTGAGCCAATAG[A/C]CTGTGTTTGCCTTTTACAGCTGAGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C22orf34 PubMed Links

Gene Details

Gene
C22orf34
Gene Name
chromosome 22 open reading frame 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289922.2 Intron NP_001276851.1

View Full Product Details