Product Details

SNP ID
rs6088068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33009454 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCAAAGCCAGGAGATCAGAAGC[A/G]GTTTCCCCCAAGGCCAAACTCAGAT
Phenotype
MIM: 614108 MIM: 613942
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPIFB2 PubMed Links

Gene Details

Gene
BPIFB2
Gene Name
BPI fold containing family B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025227.2 Intron NP_079503.1
Gene
SUN5
Gene Name
Sad1 and UNC84 domain containing 5
There are no transcripts associated with this gene.

View Full Product Details