Product Details

SNP ID
rs10283202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:85472208 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCCCTTTGTTCTGCAATGCCAAA[A/G]TAAATTTGGGCTCATTATTTTATGG
Phenotype
MIM: 611492
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CA2 PubMed Links

Gene Details

Gene
CA2
Gene Name
carbonic anhydrase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000067.2 Intron NP_000058.1
NM_001293675.1 Intron NP_001280604.1
Gene
CA3-AS1
Gene Name
CA3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details