Product Details

SNP ID
rs4590165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:83944920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAACTCCATCATCACATGACAG[A/G]ATACTTAACCCACATCTTTCAGCAT
Phenotype
MIM: 606018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDIL3 PubMed Links

Gene Details

Gene
EDIL3
Gene Name
EGF like repeats and discoidin domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278642.1 Intron NP_001265571.1
NM_005711.4 Intron NP_005702.3

View Full Product Details