Product Details
- SNP ID
-
rs5994015
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:16786059 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAAACTCTAAAACATTAGCATTGT[A/T]ACTGCATTTATGAGAAATGAGACTG
- Phenotype
-
MIM: 611674
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
XKR3
PubMed Links
Gene Details
- Gene
- XKR3
- Gene Name
- XK related 3
View Full Product Details