Product Details

SNP ID
rs6706689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:60943910 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTCCTAACTGTTCTGAGAATTAA[A/G]GTAAAAAAAGATTAATGCAAGCCTG
Phenotype
MIM: 612787
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NONOP2 PubMed Links

Gene Details

Gene
NONOP2
Gene Name
non-POU domain containing, octamer-binding pseudogene 2
There are no transcripts associated with this gene.

Gene
PUS10
Gene Name
pseudouridylate synthase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322123.1 Intron NP_001309052.1
NM_001322124.1 Intron NP_001309053.1
NM_001322127.1 Intron NP_001309056.1
NM_144709.3 Intron NP_653310.2
XM_011532568.2 Intron XP_011530870.1
XM_011532570.1 Intron XP_011530872.1
XM_011532571.1 Intron XP_011530873.1
XM_011532572.1 Intron XP_011530874.1
XM_011532573.2 Intron XP_011530875.1
XM_011532574.2 Intron XP_011530876.1
XM_011532576.2 Intron XP_011530878.1
XM_011532578.1 Intron XP_011530880.1
XM_017003428.1 Intron XP_016858917.1
XM_017003429.1 Intron XP_016858918.1

View Full Product Details