Product Details

SNP ID
rs8192609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136980213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCAGACCCCCAGGGCTGAGTTAA[A/C]GGAGAAATTCACCGCCTTCTGCAAG
Phenotype
MIM: 176803
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LCNL1 PubMed Links

Gene Details

Gene
LCNL1
Gene Name
lipocalin like 1
There are no transcripts associated with this gene.

Gene
PRR31
Gene Name
proline rich 31
There are no transcripts associated with this gene.

Gene
PTGDS
Gene Name
prostaglandin D2 synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000954.5 554 Missense Mutation AAG,ACG K160T NP_000945.3

View Full Product Details