Product Details

SNP ID
rs9584732
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:97966383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAAGCTTATCCTGCATCAATTG[A/C]GATGCTCATATAGTTTTTACGCTTT
Phenotype
MIM: 602008
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
IPO5 PubMed Links
Additional Information
For this assay, SNP(s) [rs78039536] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
IPO5
Gene Name
importin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002271.4 Intron NP_002262.3
XM_005254052.3 Intron XP_005254109.1
XM_005254053.3 Intron XP_005254110.1
XM_011521087.2 Intron XP_011519389.1
XM_011521088.2 Intron XP_011519390.1
XM_011521089.2 Intron XP_011519391.1
XM_011521090.2 Intron XP_011519392.1
XM_017020562.1 Intron XP_016876051.1

View Full Product Details