Product Details

SNP ID
rs7414521
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:88856561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAAAGCATGGCCTCCCTGACCC[A/C]TAAGATTTAAAAAGGTAAGTTTGGA
Phenotype
MIM: 189963
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GTF2B PubMed Links

Gene Details

Gene
GTF2B
Gene Name
general transcription factor IIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001514.5 Intron NP_001505.1
XM_011541299.1 Intron XP_011539601.1

View Full Product Details