Product Details

SNP ID
rs9998719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8597016 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGCTCAGTGCCAGCCGGGCACCGT[C/G]AGAGCCTTGTGATGGGTACCCAGGG
Phenotype
MIM: 603105 MIM: 606921
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPZ PubMed Links

Gene Details

Gene
CPZ
Gene Name
carboxypeptidase Z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014447.2 Intron NP_001014447.1
NM_001014448.2 Intron NP_001014448.1
NM_003652.3 Intron NP_003643.2
Gene
GPR78
Gene Name
G protein-coupled receptor 78
There are no transcripts associated with this gene.

View Full Product Details