Product Details

SNP ID
rs6119637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35670290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGGTCTAATCCAAAAGAATTC[C/T]TTGGATCTTAGACCTGAGCATCCCA
Phenotype
MIM: 604205 MIM: 603485 MIM: 607179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPNE1 PubMed Links

Gene Details

Gene
CPNE1
Gene Name
copine 1
There are no transcripts associated with this gene.

Gene
NFS1
Gene Name
NFS1 cysteine desulfurase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198989.1 Intron NP_001185918.1
NM_021100.4 Intron NP_066923.3
Gene
RBM12
Gene Name
RNA binding motif protein 12
There are no transcripts associated with this gene.

View Full Product Details