Product Details

SNP ID
rs9509371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20779914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGCTCAGCAATATGCCTAATTTG[A/G]ACATTTTAATTTTAGAAAACCAATT
Phenotype
MIM: 611449
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EEF1AKMT1 PubMed Links

Gene Details

Gene
EEF1AKMT1
Gene Name
eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1
There are no transcripts associated with this gene.

Gene
XPO4
Gene Name
exportin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022459.4 7464 UTR 3 NP_071904.4
XM_005266502.1 7464 UTR 3 XP_005266559.1
XM_005266503.3 7464 UTR 3 XP_005266560.1
XM_005266504.3 7464 UTR 3 XP_005266561.1
XM_011535197.1 7464 UTR 3 XP_011533499.1
XM_011535198.1 7464 UTR 3 XP_011533500.1
XM_011535199.1 7464 UTR 3 XP_011533501.1
XM_011535200.2 7464 UTR 3 XP_011533502.1
XM_017020704.1 7464 UTR 3 XP_016876193.1
XM_017020705.1 7464 UTR 3 XP_016876194.1
XM_017020706.1 7464 UTR 3 XP_016876195.1

View Full Product Details