Product Details

SNP ID
rs9853063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98571356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATTGAGAAATAGTTTATTTTC[A/G]TTTTTCCTTCTTTATCAATATAAAT
Phenotype
MIM: 612732
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPOX PubMed Links

Gene Details

Gene
CPOX
Gene Name
coproporphyrinogen oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000097.5 Intron NP_000088.3
XM_005247125.4 Intron XP_005247182.1

View Full Product Details