Product Details

SNP ID
rs9362013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83860942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAAAGGAAACTGGCTAAGAAAT[A/G]TTGCCCTAAATTGCGTAGGCTTGTG
Phenotype
MIM: 608343 MIM: 609891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYB5R4 PubMed Links

Gene Details

Gene
CYB5R4
Gene Name
cytochrome b5 reductase 4
There are no transcripts associated with this gene.

Gene
RIPPLY2
Gene Name
ripply transcriptional repressor 2
There are no transcripts associated with this gene.

View Full Product Details