Product Details

SNP ID
rs6630293
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:14452371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAAAATGACCCTTTCTTTGATT[C/G]TCTTTATGCTGCCTGTTTCTTCTCT
Phenotype
MIM: 305990
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GLRA2 PubMed Links

Gene Details

Gene
GLRA2
Gene Name
glycine receptor alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001118885.1 Intron NP_001112357.1
NM_001118886.1 Intron NP_001112358.1
NM_001171942.1 Intron NP_001165413.1
NM_002063.3 Intron NP_002054.1
XM_006724487.3 Intron XP_006724550.1
XM_011545495.2 Intron XP_011543797.1
XM_011545496.2 Intron XP_011543798.1
XM_017029427.1 Intron XP_016884916.1
XM_017029428.1 Intron XP_016884917.1
XM_017029429.1 Intron XP_016884918.1

View Full Product Details