Product Details

SNP ID
rs4881437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5423708 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTAGGAGGTTATTCTGTGTATC[C/T]TCTGGAAAAATAACTTGCAATCTTA
Phenotype
MIM: 606450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NET1 PubMed Links

Gene Details

Gene
NET1
Gene Name
neuroepithelial cell transforming 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001047160.2 Intron NP_001040625.1
NM_005863.4 Intron NP_005854.2

View Full Product Details