Product Details

SNP ID
rs9326430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63987006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCCTGGGATGTGAGGGGCCTGC[A/G]GGATCTGTCCCTGAGGCCTGCGACT
Phenotype
MIM: 608337
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTUB1 PubMed Links

Gene Details

Gene
OTUB1
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017670.2 Intron NP_060140.2

View Full Product Details