Product Details

SNP ID
rs10245229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139234244 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTTGAAATAGCCATCTATCACT[A/G]AATAAGATTTTAGCTCCAGATGCTG
Phenotype
MIM: 613841
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBN2 PubMed Links

Gene Details

Gene
UBN2
Gene Name
ubinuclein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173569.3 Intron NP_775840.3
XM_005250249.3 Intron XP_005250306.2
XM_006715916.3 Intron XP_006715979.2
XM_011516002.2 Intron XP_011514304.1
XM_011516003.1 Intron XP_011514305.1

View Full Product Details