Product Details

SNP ID
rs951580620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152646266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACATACCAAATTAGCAAAAGTC[A/C]AAAGTCAGACAATACCAATGTTGGT
Phenotype
MIM: 300349
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GABRQ PubMed Links

Gene Details

Gene
GABRQ
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018558.3 Intron NP_061028.3

View Full Product Details