Product Details

SNP ID
rs9850497
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12837798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTACTTAGGAAAATTCAGCTTT[A/T]TGGCTTTCATTTACCAGTAAGTGAA
Phenotype
MIM: 610403
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
CAND2 PubMed Links

Gene Details

Gene
CAND2
Gene Name
cullin associated and neddylation dissociated 2 (putative)
There are no transcripts associated with this gene.

Gene
RPL32
Gene Name
ribosomal protein L32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000994.3 Intron NP_000985.1
NM_001007073.1 Intron NP_001007074.1
NM_001007074.1 Intron NP_001007075.1
Gene
SNORA7A
Gene Name
small nucleolar RNA, H/ACA box 7A
There are no transcripts associated with this gene.

View Full Product Details