Product Details

SNP ID
rs10160213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97650603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGCTTCTGTGTCTTGCTCCTA[A/G]AAATGTAGCAATCCATTTACCTTGT
Phenotype
MIM: 609763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PI4K2A PubMed Links

Gene Details

Gene
PI4K2A
Gene Name
phosphatidylinositol 4-kinase type 2 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018425.3 Intron NP_060895.1

View Full Product Details