Product Details
- SNP ID
-
rs9332264
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:48247151 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAAATCCTAGTTTTTAAAAGTGC[A/G]AATCACTAAATTTTTTTAATGTCCC
- Phenotype
-
MIM: 603904
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ITM2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7989786] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ITM2B
- Gene Name
- integral membrane protein 2B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021999.4 |
|
Intron |
|
|
NP_068839.1 |
View Full Product Details