Product Details

SNP ID
rs10120745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:68537882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAGTCCATCCACATCCTTGCTCC[A/C]TTTCTCGCATGGCATCATCCCCTCA
Phenotype
MIM: 600981
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01506 PubMed Links

Gene Details

Gene
LINC01506
Gene Name
long intergenic non-protein coding RNA 1506
There are no transcripts associated with this gene.

Gene
LOC105376072
Gene Name
uncharacterized LOC105376072
There are no transcripts associated with this gene.

Gene
PGM5
Gene Name
phosphoglucomutase 5
There are no transcripts associated with this gene.

Gene
TMEM252
Gene Name
transmembrane protein 252
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153237.1 Intron NP_694969.1

View Full Product Details