Product Details

SNP ID
rs9904586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43803793 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTCGGAATCTCTGGGAGTAGAAC[C/T]GGGGACTCTGTGTTGAACAGCCTCA
Phenotype
MIM: 601114
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPP3 PubMed Links

Gene Details

Gene
MPP3
Gene Name
membrane palmitoylated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001932.4 Intron NP_001923.2
XM_006721915.2 Intron XP_006721978.1
XM_006721916.3 Intron XP_006721979.1
XM_011524828.2 Intron XP_011523130.1
XM_017024655.1 Intron XP_016880144.1
XM_017024656.1 Intron XP_016880145.1
XM_017024657.1 Intron XP_016880146.1
XM_017024658.1 Intron XP_016880147.1
XM_017024659.1 Intron XP_016880148.1
XM_017024660.1 Intron XP_016880149.1

View Full Product Details