Product Details

SNP ID
rs9654588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36396146 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATAGCTATACAGATCAGCGGAAC[A/C]GAATTGAGAGTACAGAAAGAAACCC
Phenotype
MIM: 605255
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ETV7 PubMed Links

Gene Details

Gene
ETV7
Gene Name
ETS variant 7
There are no transcripts associated with this gene.

Gene
PXT1
Gene Name
peroxisomal, testis specific 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152990.3 Intron NP_694535.2
XM_011514400.2 Intron XP_011512702.1

View Full Product Details