Product Details

SNP ID
rs3849967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73723608 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGAGCTCTCCAGCAACAGTGGA[C/T]CAGCTCAAAGAATAAGTGGAATATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMS1P4 PubMed Links

Gene Details

Gene
BMS1P4
Gene Name
BMS1, ribosome biogenesis factor pseudogene 4
There are no transcripts associated with this gene.

Gene
GLUD1P3
Gene Name
glutamate dehydrogenase 1 pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details