Product Details

SNP ID
rs6104476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37299467 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTTTGAAAATGGGTGTGTAATTA[A/G]CATCTCAATTCAAACTAGCTGCGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MANBAL PubMed Links

Gene Details

Gene
MANBAL
Gene Name
mannosidase beta like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003897.1 Intron NP_001003897.1
NM_022077.3 Intron NP_071360.1
XM_017028006.1 Intron XP_016883495.1
XM_017028007.1 Intron XP_016883496.1

View Full Product Details