Product Details
- SNP ID
-
rs6010133
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50515095 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAACAGAAGGCTGGAATTACACTA[A/T]TGCTGCTCACACAGTTCACTAAGGG
- Phenotype
-
MIM: 611230
MIM: 604272
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LMF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150184570] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LMF2
- Gene Name
- lipase maturation factor 2
There are no transcripts associated with this gene.
- Gene
- NCAPH2
- Gene Name
- non-SMC condensin II complex subunit H2
- Gene
- SCO2
- Gene Name
- SCO2 cytochrome c oxidase assembly protein
There are no transcripts associated with this gene.
View Full Product Details