Product Details

SNP ID
rs10256672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45163538 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCTGCCAGCAGCCAGAGGGTCGT[A/G]GAGCAGCAGCTCATGGGTGGGACAA
Phenotype
MIM: 605155
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAMP3 PubMed Links

Gene Details

Gene
RAMP3
Gene Name
receptor (G protein-coupled) activity modifying protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005856.2 Intron NP_005847.1
XM_006715631.3 Intron XP_006715694.1
XM_017011666.1 Intron XP_016867155.1

View Full Product Details