Product Details
- SNP ID
-
rs5765956
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:44754779 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACACTGTAAACGATGGAGTTAAAGG[C/T]GGCTTCGGTTAAAGAGACGAGAGCA
- Phenotype
-
MIM: 609405
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGAP8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80144674] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGAP8
- Gene Name
- Rho GTPase activating protein 8
- Gene
- PRR5-ARHGAP8
- Gene Name
- PRR5-ARHGAP8 readthrough
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181334.5 |
|
Intron |
|
|
NP_851851.3 |
View Full Product Details