Product Details

SNP ID
rs9861949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:170048471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTATGTCTTTATAAATGTTTT[C/T]AATGACTGCATGCTATTTTAGGCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR160 PubMed Links

Gene Details

Gene
GPR160
Gene Name
G protein-coupled receptor 160
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014373.2 Intron NP_055188.1
XM_005247346.3 Intron XP_005247403.1
XM_005247347.3 Intron XP_005247404.1
XM_006713582.3 Intron XP_006713645.1
XM_011512678.2 Intron XP_011510980.1
XM_017006161.1 Intron XP_016861650.1
XM_017006162.1 Intron XP_016861651.1
XM_017006163.1 Intron XP_016861652.1

View Full Product Details