Product Details
- SNP ID
-
rs6014573
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:56004403 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGTCCTTTAAGGAGCTCATGCAG[A/C]ACCCTTTACCCTACTCTCCTCCGCC
- Phenotype
-
MIM: 615029
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CBLN4
PubMed Links
Gene Details
- Gene
- CBLN4
- Gene Name
- cerebellin 4 precursor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080617.5 |
1070 |
UTR 5 |
|
|
NP_542184.1 |
View Full Product Details