Product Details

SNP ID
rs5955055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:143509529 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATAAGTCCTGTTCCCCTTATTCT[C/T]TCATCTTCTCCAGCCCTGGGCTAAC
Phenotype
MIM: 300666
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPANXN3 PubMed Links

Gene Details

Gene
SPANXN3
Gene Name
SPANX family member N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009609.3 Intron NP_001009609.2
XM_017029264.1 Intron XP_016884753.1

View Full Product Details