Product Details

SNP ID
rs6629158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:16612488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAAGTATTTGATGACAATAATGA[C/T]AACCAGAAGAATTAAAAGTGAATAA
Phenotype
MIM: 300380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTPS2 PubMed Links

Gene Details

Gene
CTPS2
Gene Name
CTP synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144002.1 Intron NP_001137474.1
NM_019857.4 Intron NP_062831.3
NM_175859.2 Intron NP_787055.1
XM_005274562.3 Intron XP_005274619.1
XM_006724503.3 Intron XP_006724566.1
XM_011545545.2 Intron XP_011543847.1

View Full Product Details