Product Details

SNP ID
rs12307010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:39543790 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTTAATACTTTATTTATGGATTC[A/T]TTCAGTCATTCTGTTACTTATTCCA
Phenotype
MIM: 601081
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCD2 PubMed Links

Gene Details

Gene
ABCD2
Gene Name
ATP binding cassette subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005164.3 Intron NP_005155.1
XM_011538027.2 Intron XP_011536329.1
XM_017018992.1 Intron XP_016874481.1
XM_017018993.1 Intron XP_016874482.1
XM_017018994.1 Intron XP_016874483.1

View Full Product Details