Product Details

SNP ID
rs12081467
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:248851876 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGTCCTCTTGCTATGATAATCCC[A/G]TGATTCCAGGCCAGTCCACTGGCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF672 PubMed Links

Gene Details

Gene
ZNF672
Gene Name
zinc finger protein 672
There are no transcripts associated with this gene.

Gene
ZNF692
Gene Name
zinc finger protein 692
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136036.2 Intron NP_001129508.1
NM_001193328.1 Intron NP_001180257.1
NM_017865.3 Intron NP_060335.2
XM_011544220.1 Intron XP_011542522.1
XM_011544221.1 Intron XP_011542523.1
XM_011544222.1 Intron XP_011542524.1
XM_011544223.2 Intron XP_011542525.1
XM_011544227.1 Intron XP_011542529.1
XM_017001698.1 Intron XP_016857187.1
XM_017001699.1 Intron XP_016857188.1
XM_017001700.1 Intron XP_016857189.1
XM_017001701.1 Intron XP_016857190.1
XM_017001702.1 Intron XP_016857191.1
XM_017001703.1 Intron XP_016857192.1
XM_017001704.1 Intron XP_016857193.1
XM_017001705.1 Intron XP_016857194.1

View Full Product Details