Product Details

SNP ID
rs12327336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5889835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTACAGGGGTCGGGAAATGAGG[G/T]ATTAAAAAAAAAAGTCGGAGTAAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM200C PubMed Links

Gene Details

Gene
TMEM200C
Gene Name
transmembrane protein 200C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080209.1 8270 Intron NP_001073678.1
XM_005258130.4 8270 UTR 3 XP_005258187.1
XM_011525729.2 8270 UTR 3 XP_011524031.1

View Full Product Details