Product Details
- SNP ID
-
rs13130564
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:183662077 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGTGCCTTACCTGAATTCTCTGT[A/G]ATATGCTGGTATAGATTATCTTTTA
- Phenotype
-
MIM: 614138
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RWDD4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116272529,rs76928504] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RWDD4
- Gene Name
- RWD domain containing 4
There are no transcripts associated with this gene.
- Gene
- TRAPPC11
- Gene Name
- trafficking protein particle complex 11
View Full Product Details