Product Details
- SNP ID
-
rs4279261
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:185457636 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTCTTGACAGTATAAAATATATAC[C/T]TAATTCCAATTTCTAATTTTAAAAA
- Phenotype
-
MIM: 609488
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C4orf47
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73873459] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C4orf47
- Gene Name
- chromosome 4 open reading frame 47
There are no transcripts associated with this gene.
- Gene
- CCDC110
- Gene Name
- coiled-coil domain containing 110
View Full Product Details