Product Details

SNP ID
rs6880051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:124638632 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTTAATTCCATTCAAATGTGGA[A/C]TTCAATTCCAGAGAAAGTGAATAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF608 PubMed Links

Gene Details

Gene
ZNF608
Gene Name
zinc finger protein 608
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020747.2 5764 Intron NP_065798.2
XM_005272037.2 5764 Intron XP_005272094.1
XM_005272038.1 5764 Intron XP_005272095.1
XM_005272039.2 5764 Intron XP_005272096.1
XM_005272040.3 5764 Intron XP_005272097.1
XM_011543520.2 5764 UTR 3 XP_011541822.1
XM_011543522.2 5764 Intron XP_011541824.1
XM_011543523.2 5764 Intron XP_011541825.1
XM_017009674.1 5764 Intron XP_016865163.1

View Full Product Details