Product Details
- SNP ID
-
rs4578731
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:13219027 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGAAGACAGTGGGTTCGGTTTAGG[G/T]TAAATCGCTATCGTGGCGGAGAGGG
- Phenotype
-
MIM: 606571
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LDLRAD4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78694231] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LDLRAD4
- Gene Name
- low density lipoprotein receptor class A domain containing 4
View Full Product Details