Product Details
- SNP ID
-
rs7218312
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:60231635 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAACAGGGACCCCCCAAAGCCCAT[A/G]GGAGAGAGGCACGAAAGGTGATTTA
- Phenotype
-
MIM: 607740
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SCARNA20
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79851920] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SCARNA20
- Gene Name
- small Cajal body-specific RNA 20
There are no transcripts associated with this gene.
- Gene
- USP32
- Gene Name
- ubiquitin specific peptidase 32
View Full Product Details