Product Details

SNP ID
rs11210732
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:42847326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGCCTTTAACCTAGGCGCTTCAC[C/G]TTCATCATCTGTGATCACTAGCCCC
Phenotype
MIM: 609017
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERMAP PubMed Links

Gene Details

Gene
ERMAP
Gene Name
erythroblast membrane associated protein (Scianna blood group)
There are no transcripts associated with this gene.

Gene
ZNF691
Gene Name
zinc finger protein 691
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242739.1 651 Intron NP_001229668.1
NM_015911.3 651 Intron NP_056995.1
XM_006710671.3 651 Intron XP_006710734.1
XM_017001400.1 651 Intron XP_016856889.1
XM_017001401.1 651 UTR 5 XP_016856890.1
XM_017001402.1 651 Intron XP_016856891.1
XM_017001403.1 651 UTR 5 XP_016856892.1

View Full Product Details