Product Details

SNP ID
rs12735446
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:152879985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGTAAAGATGGACACTAACAAA[A/C]TGGACTTCCAAGTGGTGATGAGACT
Phenotype
MIM: 601148
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMCP PubMed Links

Gene Details

Gene
SMCP
Gene Name
sperm mitochondria associated cysteine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030663.2 Intron NP_109588.2

View Full Product Details