Product Details

SNP ID
rs6593921
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:205093733 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGTCTAAATCTAGTAAACTCGAG[A/G]TGGAATACTCAAAATGATGAATAAC
Phenotype
MIM: 600697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBBP5 PubMed Links

Gene Details

Gene
RBBP5
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193272.1 Intron NP_001180201.1
NM_001193273.1 Intron NP_001180202.1
NM_005057.3 Intron NP_005048.2
XM_011509833.2 Intron XP_011508135.1
Gene
TMEM81
Gene Name
transmembrane protein 81
There are no transcripts associated with this gene.

View Full Product Details