Product Details

SNP ID
rs12645193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7971415 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGGTCTTTGCAGAGGGTGGGGC[A/G]GCCTCACTGCTGAGGTGCACTGGTT
Phenotype
MIM: 612544
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABLIM2 PubMed Links

Gene Details

Gene
ABLIM2
Gene Name
actin binding LIM protein family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130083.1 Intron NP_001123555.1
NM_001130084.1 Intron NP_001123556.1
NM_001130085.1 Intron NP_001123557.1
NM_001130086.1 Intron NP_001123558.1
NM_001130087.1 Intron NP_001123559.1
NM_001130088.1 Intron NP_001123560.1
NM_001286688.1 Intron NP_001273617.1
NM_032432.4 Intron NP_115808.3
XM_005248014.4 Intron XP_005248071.1
XM_005248017.4 Intron XP_005248074.1
XM_005248018.4 Intron XP_005248075.1
XM_005248019.4 Intron XP_005248076.1
XM_005248020.4 Intron XP_005248077.1
XM_005248021.4 Intron XP_005248078.1
XM_005248022.4 Intron XP_005248079.1
XM_005248023.4 Intron XP_005248080.1
XM_005248024.4 Intron XP_005248081.1
XM_005248027.4 Intron XP_005248084.1
XM_005248028.4 Intron XP_005248085.1
XM_005248029.4 Intron XP_005248086.1
XM_005248030.4 Intron XP_005248087.1
XM_005248031.4 Intron XP_005248088.1
XM_006713923.3 Intron XP_006713986.1
XM_006713924.3 Intron XP_006713987.1
XM_006713925.3 Intron XP_006713988.1
XM_006713927.3 Intron XP_006713990.1
XM_011513586.2 Intron XP_011511888.1
XM_017008706.1 Intron XP_016864195.1
XM_017008707.1 Intron XP_016864196.1
XM_017008708.1 Intron XP_016864197.1
XM_017008709.1 Intron XP_016864198.1
XM_017008710.1 Intron XP_016864199.1
XM_017008711.1 Intron XP_016864200.1
XM_017008712.1 Intron XP_016864201.1
XM_017008713.1 Intron XP_016864202.1
XM_017008714.1 Intron XP_016864203.1
XM_017008715.1 Intron XP_016864204.1
XM_017008716.1 Intron XP_016864205.1
XM_017008717.1 Intron XP_016864206.1
XM_017008718.1 Intron XP_016864207.1
XM_017008719.1 Intron XP_016864208.1
XM_017008720.1 Intron XP_016864209.1
XM_017008721.1 Intron XP_016864210.1
XM_017008722.1 Intron XP_016864211.1
XM_017008723.1 Intron XP_016864212.1
XM_017008724.1 Intron XP_016864213.1
XM_017008725.1 Intron XP_016864214.1
XM_017008726.1 Intron XP_016864215.1
XM_017008727.1 Intron XP_016864216.1

View Full Product Details