Product Details

SNP ID
rs11626347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105430418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTAGTTGGGGTATGGCTCATGA[A/T]GCCCAAAACAATTACAATAGTAGCA
Phenotype
MIM: 603526
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MTA1 PubMed Links

Gene Details

Gene
MTA1
Gene Name
metastasis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203258.1 Intron NP_001190187.1
NM_004689.3 Intron NP_004680.2
XM_011537305.1 Intron XP_011535607.1
XM_011537306.1 Intron XP_011535608.1
XM_011537307.1 Intron XP_011535609.1
XM_011537308.2 Intron XP_011535610.1
XM_011537309.1 Intron XP_011535611.1
XM_011537310.1 Intron XP_011535612.1
XM_017021752.1 Intron XP_016877241.1
XM_017021753.1 Intron XP_016877242.1
XM_017021754.1 Intron XP_016877243.1
XM_017021755.1 Intron XP_016877244.1
XM_017021756.1 Intron XP_016877245.1
XM_017021757.1 Intron XP_016877246.1
XM_017021758.1 Intron XP_016877247.1
XM_017021759.1 Intron XP_016877248.1

View Full Product Details