Product Details

SNP ID
rs12520325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:41904427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGAGCTCGGGGATCTGGCTCA[C/G]GCCCACATACAGCAACTTAGCGAAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C5orf51 PubMed Links

Gene Details

Gene
C5orf51
Gene Name
chromosome 5 open reading frame 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175921.5 90 Missense Mutation CAC,CAG H20Q NP_787117.3
XM_005248289.4 90 Missense Mutation CAC,CAG H20Q XP_005248346.1
XM_011514032.2 90 UTR 5 XP_011512334.1

View Full Product Details